Learn More
Gibco™ Human Relaxin-2 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand:  Gibco™ 130-15-100UG
 View more versions of this product
Product Code. 17831483
  • 606.00 € / 100µg
Explore more special offers
Description
Description
Relaxin 1 (also referred to as Relaxin or RLXH1) is a peptide hormone produced by the corpora lutea of the ovary during pregnancy in many mammalian species, including human. The secretion of the hormone into the blood stream just before parturition results in a marked softening and lengthening of the pubic symphysis and a softening of the cervix, which facilitates the birth process. By inhibiting uterine contractions, Relaxin 1 may influence the timing of parturition. Like insulin, Relaxin 1 consists of two peptide chains, A and B, covalently linked by disulfide bonds. By further analogy to insulin, the two peptides are synthesized as a single-chain precursor polypeptide with the B chain at the amino-terminus. The gene that encodes the human Relaxin 1 protein maps to chromosome 9. Relaxin 2, a related protein, is selectively expressed in the ovary during pregnancy. The gene that encodes the human Relaxin 2 protein also maps to chromosome 9.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P04090
protein with no preservative
6
100 μg
-20°C
<1 EU/μg
RLN2
Unconjugated
A-Chain: QLYSALANKC CHVGCTKRSL ARFC B-Chain: DSWMEEVIKL CGRELVRAQI AICGMSTWS
≥ 98% by SDS-PAGE gel and HPLC analyses.
Functional Assay
6019
Human Relaxin-2
E. coli
RUO
bA12D24.1.1; bA12D24.1.2; H2; H2-preprorelaxin; H2-RLX; Prorelaxin H2; relaxin 2; Relaxin A chain; Relaxin B chain; relaxin H2; relaxin, ovarian, of pregnancy; RLN2; RLX H2-RLX; RLXH2
Human
Recombinant
Lyophilized