missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RHAG (aa 27-52) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (35%), Rat (35%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63110 (PA5-63110. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Associated with rhesus blood group antigen expression. May be part of an oligomeric complex which is likely to have a transport or channel function in the erythrocyte membrane. {ECO:0000269; PubMed:11062476, ECO:0000269; PubMed:11861637}.
Specifications
Specifications
| Accession Number | Q02094 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 6005 |
| Name | Human RHAG (aa 27-52) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | Ammonium transporter Rh type A; CD241; Erythrocyte membrane glycoprotein Rh50; erythrocyte plasma membrane 50 kDa glycoprotein; OHS; OHST; Rh 50 glycoprotein; rh family type A glycoprotein; rh type A glycoprotein; RH2; RH50; Rh50A; Rh50GP; RHAG; Rh-associated glycoprotein; Rhesus associated polypeptide, 50-KD; rhesus blood group family type A glycoprotein; Rhesus blood group-associated ammonia channel; Rhesus blood group-associated glycoprotein; SLC42A1 |
| Common Name | RHAG |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction