Learn More
Invitrogen™ Human RTCB (aa 340-499) Control Fragment Recombinant Protein
Recombinant Protein
Brand:  Invitrogen™ RP91392
176.67 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Product Code. 30200510
  • 265.00 € / 100µL
Explore more special offers
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Description
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibodies, PA5-64867 (PA5-64867, PA5-51512 (PA5-51512. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Human RTCB (aa 340-499) Control Fragment Recombinant Protein > 100 μL; Unlabeled