Learn More
R&D Systems™ Human S100B DuoSet ELISA
Human S100B DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand:  R&D Systems™ DY1820-05
Product Code. 30000584
  • 468.00 € / Pack of 5
Explore more special offers
Includes: Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Description
Description
The Human S100B DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human S100B.
Specifications
Specifications
1 Kit (for 5 Plates)
Biotin
ELISA
beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta
Human
S100B
6285
Store the unopened product at 2°C to 8°C. Do not use past expiration date.