missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Serpin G1/C1 Inhibitor (Plasma) Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Inhibition Activity

Brand:  R&D Systems™ 2488-PI-200

317.92 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18327427

  • 416.00 € / 200µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Human Serpin G1/C1 Inhibitor (Plasma) Protein is derived from Human Plasma. The Human Serpin G1/C1 Inhibitor (Plasma) Protein has been validated for the following applications: Inhibition Activity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Tris and NaCl.
Serpin G1/C1 Inhibitor
Human plasma-derived Serpin G1/C1 Inhibitor protein The human plasma used for the isolation of this product were certified by the supplier to be HIV-1 and HBsAg negative at the time of shipment. Human blood products should always be treated in accordance with universal handling precautions.
Unconjugated
710
200 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers