Learn More
BD Human Soluble Protein Master Buffer Kit
Along with Human soluble protein flex set system buffer kit employs particles with discrete fluorescence intensities to detect soluble analytes at very low concentrations
Brand:  BD 558265
 View more versions of this product
Product Code. 15849268
  • 2085.00 € / 1 set
Explore more special offers
Includes:
  • Assay Diluent
  • Capture Bead Diluent
  • Detection Reagent Diluent
  • Capture Bead Diluent for Serum/Plasma
  • Wash Buffer
  • Instrument Setup Bead A1
  • Instrument Setup Bead A9
  • Instrument Setup Bead F1
  • Instrument Setup Bead F9
  • PE Instrument Setup Bead F1
  • PE Positive Control Detector
Description
Description
Buffers and instrument setup reagents provided in this kit have been optimized for analysis of analytes in tissue culture supernatants, plasma, and serum samples.
  • Provides sufficient reagents for the quantitative analysis of 100 samples and 10 instrument setup procedures or 500 samples and 10 instrument setup procedures
  • Has been optimized for use with the CBA Human Soluble Protein Flex Sets
  • Working assay range for most analytes in this system is 10 to 2,500pg/mL
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2°C to 8°C
500
Cytometric Bead Array (CBA) Assay
1 bottle 150mL assay diluent, 1 bottle 30mL capture bead diluent, 1 bottle 30mL detection reagent diluent, 1 bottle 30mL capture bead diluent for Serum/Plasma, 1 bottle 650mL wash buffer, 1 vial 0.25 mL instrument setup bead A1, 1 vial 0.25mL instrument