Learn More
Invitrogen™ Human SRC1 (aa 8-24) Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP044
Product Code. 11564700
  • 167.00 € / 50µg
Explore more special offers
Description
Description
Human SRC-1 (residues 8-24) Synthetic Peptide, PEP-044, from Invitrogen.
PEP-044 is a 17 amino acid synthetic peptide whose sequence corresponds to the N-terminal residues 8-24 of human SRC-1. The sequence of this peptide is (amino to carboxy terminus): S8-S-D-P-A-N-P-D-S-H-K-R-K-G-S-P-C24This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and SRC-1, catalog # PA1-840. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
8648
purified
Human SRC1 (aa 8-24)
-20°C. Avoid freeze-thaw cycles.
Lyophilized
Control
Yes
Peptide
RUO - research use only
Human
Q15788
0.5 mg/mL
Synthetic Peptide
peptide with no preservative
NCOA1
No preservative
50 μg
S8 - S - D - P - A - N - P - D - S - H - K - R - K - G - S - P - C24