Learn More
Gibco™ Human TGF-beta 1 Recombinant Protein, PeproTech®
Recombinant Protein
83.75 € - 5380.00 €
Specifications
Accession Number | P01137 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 7040 |
Name | Human TGF-beta 1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
17861433
|
Gibco™
100-21C-100UG |
100 μg |
1078.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
17821343
|
Gibco™
100-21C-10UG |
10 μg |
203.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
30089692
|
Gibco™
100-21C-01M |
2 x 500 μg |
5380.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
17860933
|
Gibco™
100-21C-1MG |
2 x 500 μg | N/A | N/A | N/A | |||||
17840273
|
Gibco™
100-21C-250UG |
250 μg |
1882.00 €
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
17889892
|
Gibco™
100-21C-2UG |
2 μg |
83.75 €
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
17891433
|
Gibco™
100-21C-500UG |
500 μg |
3225.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
17881443
|
Gibco™
100-21C-50UG |
50 μg |
648.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Recombinant Human TGF-beta1 is a 25.0 kDa homo-dimer, consisting of two identical 112 amino acid polypeptide chains, each weighing 12.5 kDa and linked by a single disulfide bond. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
TGF beta-1 (TGFB1, Transforming Growth Factor Beta 1) is a polypeptide member of the transforming growth factor beta superfamily of cytokines, found almost ubiquitously in tissues. Transforming growth factor (TGF)-b is stored in the extracellular matrix as a latent complex with its pro-domain. Activation of TGF beta-1 requires the binding of aV integrin to an RGD sequence in the prodomain and exertion of force on this domain, which is held in the extracellular matrix by latent TGF-b binding proteins. Latent forms are complexes of TGF beta-1, an amino-terminal portion of the TGF-beta precursor, designated TGF-LAP (TGF-latency associated peptide), and a specific binding protein, known as LTBP. TGF beta-1 helps regulates proliferation, differentiation, adhesion, migration in many cell types. Many cells have TGF beta receptors, and the protein positively and negatively regulates many other growth factors. TGF beta-1 is cleaved into a latency-associated peptide and a mature TGF beta-1 peptide, and is found in either a latent form composed of a TGFB1 homodimer, a LAP homodimer, and a latent TGFB1-binding protein, or in an active form composed of a TGF beta-1 homodimer. The mature peptide may also form heterodimers with other TGF beta family members. The gene for TGF beta-1 is frequently upregulated in tumor cells, and mutations in this gene result in Camurati-Engelmann disease and cystic fibrosis.Specifications
P01137 | |
protein with no preservative | |
Human TGF-beta 1 | |
RUO | |
Unconjugated | |
ALDTNYCFSS TEKNCCVRQL YIDFRKDLGW KWIHEPKGYH ANFCLGPCPY IWSLDTQYSK VLALYNQHNP GASAAPCCVP QALEPLPIVY YVGRKPKVEQ LSNMIVRSCK CS | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
7040 | |
CHO cells | |
Tgfb1 | |
Recombinant | |
Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.