missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TNF alpha ELISA Kits

Measure levels of human tumor necrosis factor alpha (TNFa) in human serum, plasma and cell culture supernatant samples using 96-well plates.

Brand:  Invitrogen™ EH3TNFA2

Product Code. 10290484

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Assay TNFα (human tumor necrosis factor alpha) levels in human serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples with our Invitrgen™ Human TNF alpha ELISA Kits.

These human TNF alpha ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of tumor necrosis factor alpha (TNF-alpha, TNFα) in samples at 50 to 100μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain precoated 96-well strip plate(s) and lot-optimized reagents. The economically-priced reagent kit provides validated reagents only, requiring users to coat their own plates. The antibody-pair kit consists of the matched TNFα antibody pairs and TNFα standard only. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant human TNFα (human tumor necrosis factor alpha)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 15.6 to 1000pg/mL

  • Sensitivity: < 2pg/mL

  • Precision: Dependent upon sample type and level (see instructions)

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3.5 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with human IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12, IL-12p40, IL-13, IL-15, IL-16, IL-17, IL-18, TNFβ, Eotaxin, RANTES, GROα, GROβ, MCP-1, MCP-2, MCP-3, MCP-4, VEGF, GCSF, GMCSF, MIP-1α, MIP-1β, TGFβ, IFNγ, IFNα; mouse TNFα; rat TNFα; and bovine TNFα. TNFα Receptor types 1 and 2 tested at 40mg/mL did not interfere in this assay. Recombinant pig TNFα cross-reacted at 100%

Assay range and other values listed above are for the pre-coated plate kit; see product documentation for values specific to the reagent kit and antibody-pair kit.

Includes:Complete Kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates. The Reagent Kit (previously called a “Screening Set”) contains matched antibodies, standard and detection reagents but no coated plates. The Antibody Pair Kit (previously called a “MiniKit”) contains only matched antibodies and standard (no plates or detection reagents).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

15.6 to 1000pg/mL
Human TNF alpha ELISA Kit
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
2 x 96 microplate wells
Absorbance at 450nm minus absorbance at 550nm
2 Plate Kit
Natural and recombinant human TNFa (human tumor necrosis factor alpha)
<2pg/mL
Colorimetric ELISA (TMB substrate) with Plates
No significant cross-reaction with human IL-1a, IL-1ß, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12, IL-12p40, IL-13, IL-15, IL-16, IL-17, IL-18, TNFß, Eotaxin, RANTES, GROa, GROß, MCP-1, MCP-2, MCP-3, MCP-4, VEGF, GCSF, GMCSF, MIP-1a, MIP
Lyophilized Recombinant Human TNF alpha Standard, 4 vials, TMB Substrate Solution, 2 x 13mL, Streptavidin-HRP Reagent, 2 x 14mL, Sample Diluent, 2 x 13mL, Biotinylated Antibody Reagent, 2 x 13mL, Anti-Human TNF alpha Precoated 96-well Strip Plate, 2 ea, Adhesive Plate Covers, 12 ea, 30X Wash Buffer, 2 x 50mL, Stop Solution, 2 x 13mL
Dependent upon sample type and level (see instructions)
50 μL
3.5 hr.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.