Learn More
Invitrogen™ Human TRPC6 Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP0392
Product Code. 15296599
  • 130.00 € / 50µg
Explore more special offers
Description
Description
TRPC6 Synthetic Peptide, PEP-0392, from Invitrogen.
Trpc6 forms a receptor-activated calcium channel in the cell membrane. The Trpc6 channel is activated by diacylglycerol and is thought to be under the control of a phosphatidylinositol second messenger system. Activation of the Trpc6 channel occurs independently of protein kinase C and is not triggered by low levels of intracellular calcium. Defects in the Trpc6 gene are a cause of focal segmental glomerulosclerosis 2 (FSGS2). Further, Trpc6 is a member of the mammalian transient receptor potential (TRP) superfamily can be divided into three major families including the “canonical TRP” (TRPC) family. The seven members of this family share the activation through PLC-coupled receptors and have been suggested to be components of receptor-regulated cation channels in different cell types. Furthermore, the members of the TRPC6/6/7 subfamily can be activated by diacylglycerol (DAG) analogs, suggesting a possible mechanism of activation of these channels by PLC-coupled receptors. When expressed in transfected cells, Trpc6 acts as a non-selective store-independent receptor-activated cation channel. Trpc6 is activated by DAG in a PKC-independent manner and is insensitive to IP3 activation. There is increasing evidence that Trpc6 encodes endogenous DAG-activated receptor-operated cation channels in vivo. Diseases associated with TRPC6 include Glomerulosclerosis, Focal Segmental, 2 and Focal Segmental Glomerulosclerosis. This peptide corresponds to 14 amino acids near the carboxy terminus of human TRPC6.PEP-0392 can be used as a blocking peptide with polyclonal antibody PA5-20256.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7225
purified
Human TRPC6
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q9Y210
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
TRPC6
0.02% sodium azide
50 μg
PBS