Learn More
Novus Biologicals™ Human Universal Protein Whole Tissue Lysate (Adult Whole Normal)
Brand:  Novus Biologicals™ NB820-89437
Product Code. 18233364
  • 412.00 € / Pack of 2
Explore more special offers
Description
Description
The Human Universal Protein Whole Tissue Lysate (Adult Whole Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
Western Blot
2 Vials
Universal Protein Lysate: Human Normal Tissues
Normal
Human
Human Universal Protein Whole Tissue Lysate (Adult Normal)
Adult
Universal
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Whole
Human
5 mg/mL
-80°C. Avoid freeze-thaw cycles.
Native