Learn More
Novus Biologicals™ Human Vagina Whole Tissue Lysate (Adult Whole Normal)
Brand:  Novus Biologicals™ NB820-59278
Product Code. 18204255
  • 319.00 € / 1mg
Explore more special offers
Description
Description
The Human Vagina Whole Tissue Lysate (Adult Whole Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
Immunoprecipitation, Western Blot
1 mg
Total Protein - Human Adult Normal Tissue: Vagina
Normal
Human
Human Vagina Whole Tissue Lysate (Adult Normal)
Adult
Reproductive
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Whole
Human
5 mg/mL
-80°C. Avoid freeze-thaw cycles.
Native