missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human XL Cytokine Luminex™ Performance Panel Premixed Kit

1360.00 € - 8275.00 €

Specifications

Species Human
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
View More Specs

Products 45
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30001034
View Documents
R&D Systems™
FCSTM18B-23
23 Plex
5575.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30001033
View Documents
R&D Systems™
FCSTM18B-24
24 Plex
5710.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30001030
View Documents
R&D Systems™
FCSTM18B-28
28 Plex
6250.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30001019
View Documents
R&D Systems™
FCSTM18B-39
39 Plex
7735.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30001017
View Documents
R&D Systems™
FCSTM18B-17
17 Plex
4570.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30001001
View Documents
R&D Systems™
FCSTM18B-05
5 Plex
2040.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
18722587
View Documents
R&D Systems™
FCSTM18B-31
31 Plex
6655.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000992
View Documents
R&D Systems™
FCSTM18B-41
41 Plex
7950.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000983
View Documents
R&D Systems™
FCSTM18B-18
18 Plex
4770.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000979
View Documents
R&D Systems™
FCSTM18B-43
43 Plex
8115.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000978
View Documents
R&D Systems™
FCSTM18B-11
11 Plex
3370.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000957
View Documents
R&D Systems™
FCSTM18B-27
27 Plex
6115.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000955
View Documents
R&D Systems™
FCSTM18B-37
37 Plex
7465.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000937
View Documents
R&D Systems™
FCSTM18B-45
45 Plex
8275.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
18793377
View Documents
R&D Systems™
FCSTM18B-42
42 Plex
8030.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000922
View Documents
R&D Systems™
FCSTM18B-44
44 Plex
8195.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000920
View Documents
R&D Systems™
FCSTM18B-06
6 Plex
2265.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
18754277
View Documents
R&D Systems™
FCSTM18B-03
3 Plex
1585.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000908
View Documents
R&D Systems™
FCSTM18B-30
30 Plex
6520.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000906
View Documents
R&D Systems™
FCSTM18B-38
38 Plex
7600.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000899
View Documents
R&D Systems™
FCSTM18B-15
15 Plex
4170.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
18732627
View Documents
R&D Systems™
FCSTM18B-10
10 Plex
3170.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
18702377
View Documents
R&D Systems™
FCSTM18B-33
33 Plex
6925.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
18704447
View Documents
R&D Systems™
FCSTM18B-08
8 Plex
2720.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
18782497
View Documents
R&D Systems™
FCSTM18B-36
36 Plex
7330.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000858
View Documents
R&D Systems™
FCSTM18B-16
16 Plex
4370.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
18700617
View Documents
R&D Systems™
FCSTM18B-07
7 Plex
2490.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000854
View Documents
R&D Systems™
FCSTM18B-19
19 Plex
4970.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000841
View Documents
R&D Systems™
FCSTM18B-20
20 Plex
5170.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000840
View Documents
R&D Systems™
FCSTM18B-14
14 Plex
3970.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000832
View Documents
R&D Systems™
FCSTM18B-04
4 Plex
1810.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000828
View Documents
R&D Systems™
FCSTM18B-35
35 Plex
7195.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
18792687
View Documents
R&D Systems™
FCSTM18B-09
9 Plex
2945.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000813
View Documents
R&D Systems™
FCSTM18B-02
2 Plex
1360.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000796
View Documents
R&D Systems™
FCSTM18B-26
26 Plex
5980.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000786
View Documents
R&D Systems™
FCSTM18B-46
46 Plex
6695.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000770
View Documents
R&D Systems™
FCSTM18B-29
29 Plex
6385.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000761
View Documents
R&D Systems™
FCSTM18B-25
25 Plex
5845.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000757
View Documents
R&D Systems™
FCSTM18B-12
12 Plex
3570.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000755
View Documents
R&D Systems™
FCSTM18B-22
22 Plex
5440.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000745
View Documents
R&D Systems™
FCSTM18B-34
34 Plex
7060.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000743
View Documents
R&D Systems™
FCSTM18B-32
32 Plex
6790.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000742
View Documents
R&D Systems™
FCSTM18B-40
40 Plex
7870.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000738
View Documents
R&D Systems™
FCSTM18B-21
21 Plex
5305.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
18772167
View Documents
R&D Systems™
FCSTM18B-13
13 Plex
3770.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Human
ELISA
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.