Learn More
Proteintech™ HumanKine™ Recombinant Human Sonic Hedgehog (SHH) Protein
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
Brand:  Proteintech™ HZ-1306-100UG
 View more versions of this product
Product Code. 16881416
  • 792.00 € / 100µg
Explore more special offers
Description
Description
  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Specifications
Specifications
6469
100 μg
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile water containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Typically ≤ 350 ng/mL EC50
Lyophilized
20 to 24 kDa
Human
Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Human embryonic kidney 293 (HEK293) human cell expression system
> 90%