missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ HumanKine™ Recombinant Human Thrombopoietin Protein

HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells

137.00 € - 2994.00 €

Specifications

Gene ID (Entrez) 7066
Molecular Weight (g/mol) 80 to 85 kDa, monomer, glycosylated
Species Human
Reconstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Content And Storage Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16871386
View Documents Promotion Details
Proteintech™
HZ-1248-10UG
10 μg
137.00 €
10µg
Please to purchase this item. Need a web account? Register with us today!
16861386
View Documents Promotion Details
Proteintech™
HZ-1248-100UG
100 μg
594.00 €
100µg
Please to purchase this item. Need a web account? Register with us today!
16851386
View Documents Promotion Details
Proteintech™
HZ-1248-1000UG
1000 μg
2994.00 €
1000µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Specifications

Specifications

7066
Human
Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Human embryonic kidney 293 (HEK293) human cell expression system
> 95%
80 to 85 kDa, monomer, glycosylated
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Typically ≤ 5 ng/mL EC50
Lyophilized
Videos
SDS
Documents

Documents

Product Certifications