missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HuMEC Basal Serum-Free Medium (1X)
GREENER_CHOICE

HuMEC Basal Serum-Free Medium (1X)

Brand:  Gibco™ 12753018


Product Code. 10120413

  • 176.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

HuMEC Basal Serum-Free Medium is designed to work with HuMEC Supplement Kit (SKU# 12755013). Together, these products are engineered specifically for the culture of human mammary epithelial cells (SKU# A10565). This translates to greater experimental validity, more reproducible results, and reduction of lot-to-lot variability and aberrations that can occur with the addition of serum. Human mammary epithelial cells cultured in HuMEC Medium have gene expression profiles consistent with normal mammary epithelial cells, as tested on a cDNA microarray.

Applications:
HuMEC Medium is optimized specifically for the growth of human mammary epithelial cells, and has been performance tested in a growth assay using normal human mammary epithelial cells. HuMEC Medium promotes superior growth of normal human mammary epithelial cells, making it ideal for researchers in breast cancer, drug discovery, and other aspects of breast biology.

Contents and Storage:
HuMEC Basal Serum Free Medium should be supplemented with HuMEC Supplement Kit. HuMEC Supplement Kit and HuMEC Basal Serum Free Media can be ordered together as HuMEC Ready Medium (SKU# 12752010). The HuMEC Supplement Kit includes 5 ml of a supplement mix containing epidermal growth factor, hydrocortisone, isoproterenol, transferrin, and insulin, and 25 mg of bovine pituitary extract. Store the basal medium at +2°C to +8°C.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Epithelial Cell (Mammary)
1 X
HuMEC Basal Serum-Free Medium
Phenol Red
Serum-free
Liquid
Sterile-filtered
500 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • N-Methyl-N-propargylbenzylamine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : Pargyline

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.