Learn More
Gibco™ HuMEC Supplement Kit
HuMEC Supplement Kit
Brand:  Gibco™ 12755013
Product Code. 10563565
  • 160.00 € / Each
Explore more special offers
Description
Description
HuMEC Supplement Kit contains growth factors and other components and is designed to work with HuMEC Basal Serum-Free Medium (12753018). Together, these products are engineered specifically for the culture of human mammary epithelial cells (A10565). This translates to greater experimental validity, more reproducible results, and reduction of lot-to-lot variability and aberrations that can occur with the addition of serum. Human mammary epithelial cells cultured in HuMEC Medium have gene expression profiles consistent with normal mammary epithelial cells, as tested on a cDNA microarray.
Applications:
HuMEC Medium is optimized specifically for the growth of human mammary epithelial cells, and has been performance tested in a growth assay using normal human mammary epithelial cells. HuMEC Medium promotes superior growth of normal human mammary epithelial cells, making it ideal for researchers in breast cancer, drug discovery, and other aspects of breast biology.
Contents and Storage:
The HuMEC Supplement Kit includes 5 ml of a supplement mix containing epidermal growth factor, hydrocortisone, isoproterenol, transferrin, and insulin, and 25 mg of bovine pituitary extract. Store the supplement mix at -5°C to -20°C.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Epithelial Cells (Mammary)
HuMEC Supplement
Sterile-filtered
Human
Mammalian Cell Culture
Serum-Free
1 Kit