missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Potent RXR antagonist (IC50 = 18 nM). Promotes white and brown pre-adipocyte differentiation into white adipocytes. Also inhibits bexarotene -induced brown adipogenic reprogramming of myoblasts.

Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 483.56 |
Percent Purity | >98% |
Chemical Name or Material | HX 531 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HX 531, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction