missing translation for 'onlineSavingsMsg'
Learn More

Hyaluronic Acid, MP Biomedicals™

Product Code. 11440112
Click to view available options
10mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Hyaluronic Acid from bovine vitreous humor

TRUSTED_SUSTAINABILITY

Specifications

CAS 9004-61-9
Color Tan
Molecular Formula C28H44N2O23
Quantity 10 mg
Synonym hyaluronic acid, dsstox_cid_26750, dsstox_rid_81875, dsstox_gsid_46750, 4xi-alpha-d-xylo-hexopyranuronosyl-1->3-3xi-2-acetylamino-2-deoxy-beta-d-ribo-hexopyranosyl-1->4-5xi-beta-d-xylo-hexopyranuronosyl-1->3-2-acetylamino-2-deoxy-d-glucopyranose
InChI Key KIUKXJAPPMFGSW-YXBJCWEESA-N
SMILES CC(=O)NC1C(C(C(OC1O)CO)O)OC2C(C(C(C(O2)C(=O)O)OC3C(C(C(C(O3)CO)O)OC4C(C(C(C(O4)C(=O)O)O)O)O)NC(=O)C)O)O
IUPAC Name (2S,4S,5R,6S)-6-[(2S,3R,5S,6R)-3-acetamido-2-[(3S,4R,5R,6R)-6-[(3R,4R,5S,6R)-3-acetamido-2,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-2-carboxy-4,5-dihydroxyoxan-3-yl]oxy-5-hydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
Molecular Weight (g/mol) 776.651
PubChem CID 24728612
Physical Form Crystalline Powder
Chemical Name or Material Hyaluronic Acid
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : hyaluronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.