missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Hyaluronidase

Product Code. 17199271
Encompass_Preferred
Click to view available options
Quantity:
300,000 U
50,000 U
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17199271

Brand: STEMCELL Technologies 07461

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For hydrolysis of hyaluronic acid

  • Hyaluronidase is a polysaccharidase consisting of a single polypeptide chain of 450 amino acid residues and contains four disulfide bonds
  • Hyaluronidase is glycosylated and contains 5% mannose and 2.17% glucosamine (Borders & Raftery)
  • Hyaluronidase cleaves endo-N-acetlyhexosaminic bonds in hyaluronic acid and chondroitin sulfate A and C to tetrasaccharide residues, as hyaluronic acid and chondroitin sulfate are often found in connective tissues
  • Hyaluronidase is frequently used with other proteases such as collagenase

Specifications

For Use With (Application) Hydrolysis of Hyaluronic Acid
Quantity 50,000 U
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.