Learn More
Cytiva Hybond™ Blotting Paper
Use with Hybond™ blotting membranes. Cytiva Life Sciences™ Hybond™ Blotting Paper is pure cellulose grade and perfect for gel electrophoresis applications.
Brand:  Cytiva RPN6101M
81.17 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
Product Code. 11750094
  • 92.97 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For gel electrophoresis
  • Used with Hybond blotting membranes
  • Pure cellulose grade
  • Medium wet strength
  • Smooth surface
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gel Electrophoresis, Hybond Blotting Membranes
0.01 in.
7.9 x 7.9 in. (20 x 20 cm)
20 cm
20 cm
Hybond Blotting Paper
0.03 cm
7.9 in.
7.9 in.