missing translation for 'onlineSavingsMsg'
Learn More

UVP Hybridization Oven Bottles and Accessories

Product Code. 11778231
Click to view available options
Product Type:
Acrylic Carousel (for upper chamber)
Carousel Kit
Hybridization Bottle (15 cm L)
Hybridization Bottle (30 cm L)
Rocker Tray
Material:
Borosilicate Glass
Stainless Steel
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11778231

Brand: UVP 07019402

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Closure Material Polypropylene, PTFE
Color clear
For Use With (Application) DNA, RNA, or protein identification
For Use With (Equipment) UVP hybridization ovens
Length (Metric) 30 cm
Material Borosilicate Glass
Product Type Hybridization Bottle (30 cm L)
Volume (Metric) 285 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.