Learn More
UVP Hybridization Oven Bottles and Accessories
Brand:  UVP 76-0075-01
 View more versions of this product
Product Code. 12863396
  • 289.00 € / Each
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Polypropylene, PTFE
UVP hybridization ovens
Borosilicate Glass
DNA, RNA, or protein identification
115 mm
Carousel Kit