missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Techne™ Hybridization Tube Holder
Expand the capabilities of your incubator with these parts and accessories.
Brand: Techne™ 39455-28
This item is not returnable.
View return policy
Description
- Provided with clips
- Available for either 16 x 15 mL or 8 x 50 mL
- Aids in applications such as nucleic acid hybridizations such as Northern (RNA) blots/hybridizations, Southern (DNA) blots/hybridizations, Western (protein) blots, and molecular biology assays
Specifications
16 x 15mL Tubes | |
Tube Carrier/Holder | |
Northern, Southern and Western blots, molecular biology assays |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction