Learn More
Cytiva HyCell™ TransFx™-C Media
For transient transfection of CHO cells and production of a variety of recombinant proteins manufactured under cGMP in ISO-compliant facilities
Brand: Cytiva SH30941.02
65.41 EUR valid until 2024-12-31
Use promo code "21885" to get your promotional price.
Code : NEW
Additional Details : Weight : 1.00000kg
Description
- Defined hydrolosate-free ADCF cell culture media formulation
- Supports transient transfection and growth of multiple CHO cell lines
- Supports the transient expression of a variety of recombinant proteins
- Same media for transient transfection and production
- Developed through our Metabolic Pathway Design process for high transfection efficiency, high viable cell yields, and recombinant protein production
- Component traceability
- Manufactured under cGMP in ISO-compliant facility
Order Info
Available in liquid form in 1L bottles, 5 to 500L bioprocess containers or custom packaging. The media is also available as a powder in pack sizes from 5 to 1,000L for bioprocess production applications.
Specifications
2° to 8°C, away from light | |
HyCell TransFx-C liquid medium - 1 L | |
Sodium Bicarbonate | |
1 L bottle |
Liquid | |
0.2 μm Sterile-filtered | |
No Poloxamer 188 |