missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HyClone™ 1 M NaCl, Cytiva
Click to view available options
Quantity:
1 L
10 L
20 L
50 L
100 L
200 L
Packaging:
Single-use Bioprocessing Container
Description
HyClone™ NaCl solutions in single-use bags offer a convenient format for many upstream and downstream bioprocessing applications.
- Manufactured in ISO 9001 and ISO 13485 certified facility.
- Lot-specific certificate of analysis (CoA) for each lot produced.
- Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
- 0.2 μm sterile filtered.
- Provided in single-use bag containers.
- Raw material is EP, USP, and JP grade.
Specifications
Specifications
Color | Clear, Colorless |
Quantity | 50 L |
Concentration or Composition (by Analyte or Components) | 1.0 M (1.0 N) |
Packaging | Single-use Bioprocessing Container |
Product Line | HyClone™ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction