missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ AdvanceSTEM™ ES Qualified Amino Acids
Brand: Cytiva SH30853.01
17.58 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
Support growth of embyronic cell culture with HyClone™ AdvanceSTEM™ ES Qualified Amino Acids.
Specifications
2° to 8°C | |
Non-Essential Amino Acids | |
Yes |
100 X | |
100 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- MEM Non-Essential Amino Acids Solution
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P310-Immediately call a POISON CENTER/doctor/.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305-IF IN EYES:Immediately call a POISON CENTER/doctor..
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction