Learn More
Cytiva HyClone™ Alpha Calf Fraction media supplement (U.S. origin)
Promote growth with this cost-effective media supplement, designed for antibody production in hybridoma cell cultures.
Brand: Cytiva SH30212.03
Code : 84
Additional Details : Weight : 0.84000kg
Description
HyClone Alpha Calf Fraction is a cost-effective medium supplement that promotes growth and antibody production in hybridoma cell cultures. This product is produced from our bovine calf serum, using a non-ethanol proprietary process to reduce immunoglobulins and protein to levels very similar to those of fetal bovine serum (FBS). Alpha Calf Fraction typically contains less than 200 μg IgG /mL. Alpha Calf Fraction is filtered through three sequential 100 nm (0.1 µm) pore size-rated filters. Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.
Specifications
Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose | |
Without Iron | |
Sterile | |
Filtered through three sequential 100 nm (0.1 µm) pore size-rated filters |