missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Bovine Growth Serum (U.S.)
Support cell culture growth with U.S.-sourced HyClone™ Bovine Growth Serum, a superior performing fetal bovine serum (FBS) replacement, consisting of bovine calf serum supplemented with chemically-defined components.
Brand: Cytiva SH30541.03
This item is not returnable.
View return policy
Description
- Filtered through sequential 100nm (0.1μm) pore-size rated filters

Specifications
Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Insulin, Iron, Lactate Dehydrogenase, Osmolality, Percent Saturation, pH, Phosphorus, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein | |
United States | |
500 mL | |
Sequential-filtered, 100 nm | |
Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
≤ 10 mg/dL |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
Bovine Growth Serum | |
Sterile | |
None | |
≤ 10 EU/mL | |
No |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 10
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HyClone™ Bovine Growth Serum (U.S.) > 500mL
Spot an opportunity for improvement?Share a Content Correction