missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ CCM3™ Media

Brand:  Cytiva SH30061.03

Product Code. 10741274

  • 220.00 € / 10 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2° to 8°C, away from light and moisture
Powder
Glutamine
10 L
CCM3 (DPM)
Non-sterile
No Calcium Chloride, No Sodium Bicarbonate, No Phenol Red, No HEPES
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva HyClone™ CCM3™ Media > 1 x 10L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.