missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GE Healthcare Hyclone™ HyClone™ CDM4MAb Media
39.90 €
Specifications
Certifications/Compliance | cGMP |
---|---|
Content And Storage | 2 to 8°C |
Form | Liquid |
Product Type | Hyclone CDM4MAb Media |
Without Additives | No Glutamine |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10054523
|
GE Healthcare Hyclone™
SH30802.01 |
500 mL | N/A | ||||||
Description
Specifications
cGMP | |
Liquid | |
No Glutamine | |
500 mL |
2 to 8°C | |
Hyclone CDM4MAb Media | |
Production of monoclonal antAntibodies (MAbs) from hybridoma and recombinant myeloma cell lines for research, therapeutic and diagnostic purposes is rapidly growing. |