Learn More
Cytiva HyClone™ Cell Boost™ 4 Supplement
Fed-batch media supplement containing amino acids, vitamins, glucose, trace elements, and growth factors for CHO cells
Brand:  Cytiva SH31116.02
 View more versions of this product
Product Code. 16685795
  • 209.00 € / 1 L
Explore more special offers
Description
Description
  • Chemically Defined (CD), Animal-Derived Component-Free (ADCF), Protein-Free, and Hydrolysate-Free
  • Supports fed-batch cultures with amino acids, vitamins, glucose, trace elements, and growth factors
  • Developed for recombinant protein production with various cell lines including Chinese Hamster Ovary (CHO) cell line
  • Designed to support modern cell culture manufacturing processes in research and bioprocessing applications
  • Manufactured to meet cGMP manufacturing standards and QC specifications
  • Available as a ready-to-use liquid feed solution
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CHO
Cell Boost Supplement
Improving Productivity and Cell Yield in Fed-Batch Cultures
1000 mL
Liquid
Bottle
HyClone™