missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Cell Boost 5™ Supplement

Provides cell culture nutrients such as lipids, amino acids, vitamins, and growth factors. HyClone™ Cell Boost™ 5 Supplement is designed through Metabolic Pathway Design approach to support demand in the area of fed-batch cell culture and media customization.

Brand:  Cytiva SH30865.01

285.09 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.


Product Code. 11566341

  • 327.00 € / 100g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Cell Boost 5 (CN-F) powdered supplement without L-glutamine
  • Non-sterile
  • Chemically defined
  • Free of animal-derived components
  • Tested in a variety of cell lines, including CHO, Hybridoma, NS0, and HEK 293
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at 2° to 8°C.
Non-sterile
100 g
Cell Boost Supplement
Bioprocessing, fed-batch cell culture, and media customization applications
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Hazard Statement
  • EUH210-Safety data sheet available on request.

Supplemental information
  • UNKNOWN AQUATIC-Contains 71.8% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.