missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Cell Boost 7b Supplement

Supplements cell culture with concentrated amino acids. Manufactured to meet cGMP manufacturing standards and QC specifications.

Brand:  Cytiva SH31027.07

409.17 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.



 View more versions of this product

Product Code. 16457695

  • 477.00 € / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture. HyClone Cell Boost 7a and Cell Boost 7b supplements are intended for use in combination with HyClone ActiPro ActiSM media as part of the ActiPro cell culture system formulated to provide high yields of recombinant proteins in processes when using Chinese hamster ovary (CHO) cell lines.

Cell Boost 7b is available as a convenient ready-to-use liquid feed solution or as a dry powder feed to be hydrated before use.

  • Designed to support modern manufacturing processes, Cell Boost 7b supplement is a concentrated solution of amino acids with an alkaline pH. Supplement is chemically defined, animal-derived component-free (ADCF), do not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol
  • Use Cell Boost 7a and 7b together to achieve high yields of mAbs and other proteins in CHO fed-batch processes
  • For convenience, this cell culture supplement is available as a ready-to-use liquid feed solution in addition to a dry powder format
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2 to 8°C, away from light and moisture
Non-sterile
1 L
Powder
No Glutamine, No Poloxamer 188, No Insulin
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Cell Boost 7b, without Poloxamer-188, without Insulin, without L-Glutamine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : tyrosine
  • UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers