missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum (FBS), U.S. Origin
Low in antibodies and high in growth factors to support robust growth and division of cells, including embryonic stem cells.
Brand: Cytiva SH30071.03IH25-40
590.21 EUR valid until 2024-12-31
Use promo code "21878" to get your promotional price.
This item is not returnable.
View return policy
Description
- Additional traceability testing of HyClone™ Fetal Bovine Serum is performed by a collaborator, Oritain
- Low in antibodies and high in growth factors
- Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
- Each lot is subjected to virus panel testing according to 9 CFR 113.53
- Characterized FBS is assayed for many components, including gamma globulin, glucose, and IgG
- Supports robust growth and division of cells, including embryonic stem cells
- Produced using true pool technology to minimize bottle-to-bottle variability
- Complete traceability is provided back to the original source, as certified by the International Serum Industry Association (ISIA)
- Sourced in the USA
Specifications
United States | |
Fetal Bovine Serum | |
Sterile | |
Characterized | |
Virus Panel Testing According to 9 CFR 113.53 | |
HyClone™ |
Serum | |
500 mL | |
Sequential Triple-filtered, 100 nm | |
Heat Inactivated, Irradiated (25-40 kGy) | |
500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction