missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum (FBS), U.S. Origin

Product Code. 16625735
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Processing:
HI, Irr - 25 to 40 doses
HI, Irr - 30 to 45 doses
Heat Inactivated, Irradiated
Irradiated
None
Unit Size:
1 L
100mL
500mL
This item is not returnable. View return policy

Product Code. 16625735

Brand: Cytiva SH30071.02IR

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Low in antibodies and high in growth factors to support robust growth and division of cells, including embryonic stem cells.

  • Additional traceability testing of HyClone™ Fetal Bovine Serum is performed by a collaborator, Oritain
  • Low in antibodies and high in growth factors
  • Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
  • Each lot is subjected to virus panel testing according to 9 CFR 113.53
  • Characterized FBS is assayed for many components, including gamma globulin, glucose, and IgG
  • Supports robust growth and division of cells, including embryonic stem cells
  • Produced using true pool technology to minimize bottle-to-bottle variability
  • Complete traceability is provided back to the original source, as certified by the International Serum Industry Association (ISIA)
  • Sourced in the USA
TRUSTED_SUSTAINABILITY

Specifications

Country of Origin United States
Form Serum
Product Type Fetal Bovine Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential Triple-filtered, 100 nm
Grade Characterized
Processing Irradiated
Tested For Virus Panel Testing According to 9 CFR 113.53
Volume (Metric) 100 mL
Product Line HyClone
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.