missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum (New Zealand Origin)
Enhance growth of cells with New Zealand sourced Characterized FBS.
775.00 € - 1080.00 €
Specifications
Additional Assays | Progesterone,Chloride,Insulin,Inorganic Phosphorus,PTH,Glucose,Cholesterol,Copper,HDL,Magnesium,LDL,Selenium,Triglycerides,IgG,Gamma Globulin,pH,Phospholipids,Osmolality,Alkaline Phosphatase,Iron,Lactate Dehydrogenase,TIBC,SGPT,Percent Saturation,SGOT,B1,B2,B6,Niacin,Biotin,Retinol,Carotene,Tocopherol,B12,Folate,Vitamin C,1,25-dihydroxy vitamin D,Total Protein,Albumin,Blood Urea Nitrogen |
---|---|
Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C |
Sterility | Sterile |
Sterilization Method | Sequential-filtered, 100 nm |
Tested For | Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus |
Product Code | Brand | Processing | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Processing | Quantity | Price | Quantity & Availability | ||||
16271112
|
Cytiva
SH30406.02IR25-40 |
Irradiated, 25 to 40 doses | 500 mL |
1080.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10711820
|
Cytiva
SH30406.02IR |
None | 500 mL | N/A | |||||
16291112
|
Cytiva
SH30406.02IR30-45 |
Irradiated, 30 to 45 doses | 500 mL |
1080.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Specifications
Progesterone,Chloride,Insulin,Inorganic Phosphorus,PTH,Glucose,Cholesterol,Copper,HDL,Magnesium,LDL,Selenium,Triglycerides,IgG,Gamma Globulin,pH,Phospholipids,Osmolality,Alkaline Phosphatase,Iron,Lactate Dehydrogenase,TIBC,SGPT,Percent Saturation,SGOT,B1,B2,B6,Niacin,Biotin,Retinol,Carotene,Tocopherol,B12,Folate,Vitamin C,1,25-dihydroxy vitamin D,Total Protein,Albumin,Blood Urea Nitrogen | |
Sterile | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
≤ 25 mg/dL | |
USDA Inspected |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C | |
Sequential-filtered, 100 nm | |
≤ 20 EU/mL | |
Yes |