missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Cosmic Calf™ Serum, New Zealand Origin
Fortified with iron and growth promoting factors for growing a broad spectrum of cell types
Brand: Cytiva SH30413.03IR30-45
Additional Details : Weight : 1.15000kg
Description
- Sourced in New Zealand with complete traceability back to original source
- Age of animal at time of collection less than 10 days old
- Low in antibodies and high in growth factors
- Filtered through sequential 100 nm pore size-rated filters
- Virus panel testing according to 9 CFR 113.53
- Each serum lot is pooled using true pool technology to ensure uniformity and consistency between bottles
Specifications
Insulin, Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Iron, TIBC, Percent Saturation, Glucose, and IgG | |
<10 days | |
Calf Bovine Serum | |
Sequential-filtered, 100 nm | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, and Reovirus | |
≤ 25 mg/dL | |
Yes |
Store ≤-10°C | |
New Zealand | |
1000 mL | |
Irradiated (30-45 kGy) | |
≤ 25 EU/mL | |
HyClone™ |