Learn More
Cytiva HyClone™ Cosmic Calf Serum (U.S. origin)
A bovine calf serum fortified with iron and growth promoting factors for growing a broad spectrum of cell types.
Brand: Cytiva SH30087.02HI
40.85 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
HyClone Cosmic Calf serum is a high-performing bovine calf serum that has been fortified with iron and a proprietary blend of growth-promoting factors. The result is a naturally derived, high-quality engineered serum that has demonstrated excellent performance with a broad range of cell types. All components of Cosmic Calf serum are sourced in the USA and are traceable.
Specifications
Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C | |
100 mL | |
Sequential-filtered, 100 nm, Gamma-irradiated | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
FBS Replacement Serum | |
Sterile | |
Heat Inactivated | |
Yes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.