Learn More
Cytiva HyClone™ Cosmic Calf Serum (U.S. origin)
A bovine calf serum fortified with iron and growth promoting factors for growing a broad spectrum of cell types.
Brand:  Cytiva SH30087.04HI
219.45 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
 View more versions of this product
Product Code. 12694227
  • 231.00 € / 1000mL
Explore more special offers
Description
Description
HyClone Cosmic Calf serum is a high-performing bovine calf serum that has been fortified with iron and a proprietary blend of growth-promoting factors. The result is a naturally derived, high-quality engineered serum that has demonstrated excellent performance with a broad range of cell types. All components of Cosmic Calf serum are sourced in the USA and are traceable.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C
1000 mL
Sequential-filtered, 100 nm, Gamma-irradiated
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Calf Bovine Serum
Sterile
Heat Inactivated
Yes