missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Donor Adult Bovine Serum (U.S.)
Used to enhance cell culture performance; a cost-efficient alternative to Fetal Bovine Serum (FBS).
370.00 € - 391.68 €
Specifications
Certifications/Compliance | Certificate of Suitability No. R1-CEP 2001-259 |
---|---|
Country of Origin | United States |
Form | Liquid |
Product Type | Adult Bovine Serum |
Quantity | 1 L |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16635745
|
Cytiva
SH30075.05 |
1 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17296936
|
Cytiva
SH30075.05IR |
1 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16271092
|
Cytiva
SH30075.05IR30-45 |
1 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Sourced from a closed herd maintained on USDA-licensed premises
- Cattle from these closed herds are used exclusively for the production of donor bovine blood products
Specifications
Certificate of Suitability No. R1-CEP 2001-259 | |
Liquid | |
1 L | |
≤ 50 EU/mL | |
≤ 25 mg/dL |
United States | |
Adult Bovine Serum | |
Sequential Triple-filtered, 100 nm | |
No | |
HyClone™ |