missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Donor Equine Serum (U.S.)
Supplement cell culture growth with U.S. sourced, collected from selected donor herds that receive regular veterinary inspection and care.
Brand: Cytiva SH30074.03
56.34 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- Format: Liquid
Specifications
Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein | |
United States | |
Equine Serum | |
Sequential Triple-filtered, 100 nm | |
BVDV, Cytopathogenic Agents, Rabies Virus, Reovirus |
Donor Equine Serum (U.S.) | |
Liquid | |
500 mL | |
None | |
No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction