Learn More
Cytiva HyClone™ Donor Equine Serum (U.S. origin)
Supplement cell culture growth with U.S. sourced, collected from selected donor herds that receive regular veterinary inspection and care. Complete traceability back to the original source.
Brand:  Cytiva SH30074.02HI
 View more versions of this product
Product Code. 12694207
  • 33.00 € / 100mL
Explore more special offers
Description
Description
  • Donor herds are fed an enriched diet to ensure proper nutrition and are fasted prior to bleeding to reduce lipid concentration
  • The horses are bled in an enclosed facility that is specially equipped for aseptic collection
  • Coggins tests for equine infectious anemia are routinely performed on each horse
  • Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG
    Sequential-filtered, 100 nm
    BVDV,Rabies Virus,Reovirus,Cytopathogenic Agents
    100 mL
    Heat Inactivated
    Yes