Learn More
Cytiva HyClone™ Donor Equine Serum (U.S. origin)
Supplement cell culture growth with U.S. sourced, collected from selected donor herds that receive regular veterinary inspection and care. Complete traceability back to the original source.
Brand: Cytiva SH30074.03IR25-40
This item is not returnable.
View return policy
Description
Specifications
Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG | |
Sequential-filtered, 100 nm | |
BVDV, Rabies Virus, Reovirus, Cytopathogenic Agents |
500 mL | |
Irradiated (25 to 40 kGy) | |
No |