missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Donor Equine Serum, US Origin

Used to enhance cell culture performance; a cost-efficient alternative to Fetal Bovine Serum (FBS).

Brand:  Cytiva SH30074.04HI

 View more versions of this product

Product Code. 16625745

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Carefully collected from animals fed an enriched diet to ensure proper nutrition
  • To reduce lipid concentration, the equine herd is fasted and then the blood is collected in a special facility using aseptic techniques
  • Coggins tests for equine infectious anemia are performed on each horse
TRUSTED_SUSTAINABILITY
Specifications

Specifications

>6 months
Liquid
1000 mL
Heat Inactivated
Yes
HyClone™
United States
Equine Serum
Sequential Triple-filtered, 100 nm
≤ 10 EU/mL
≤ 10 mg/dL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers