Learn More
Cytiva HyClone™ Donor Equine Serum, US Origin, Irradiated 25-40 kGy, Heat-Inactivated
Complete traceability back to the original source.
Brand: Cytiva SH30074.03IH25-40
This item is not returnable.
View return policy
Description
- Tested for equine infectious anemia
- Complete traceability back to original source
- Virus panel testing according to 9 CFR 113.53
Specifications
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2°C to 8°C. | |
500 mL | |
≤ 10 mg/dL |
United States | |
Irradiated | |
Not Detected |