Learn More
Cytiva HyClone™ Donor Equine Serum, US Origin, Irradiated 30-45 kGy
Complete traceability back to the original source.
Brand:  Cytiva SH30074.04IR30-45
Product Code. 17642075
  • 149.00 € / 1 L
Explore more special offers
Description
Description
  • Tested for equine infectious anemia
  • Complete traceability back to original source
  • Virus panel testing according to 9 CFR 113.53
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2°C to 8°C.
1000 mL
≤ 10 EU/mL
Not Detected
United States
Irradiated
≤ 10 mg/dL