missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Dulbecco's Modified Eagles Medium
Designed to support cell culture growth and biomanufacturing processes.
17.50 € - 188.00 €
Specifications
Certifications/Compliance | ISO 9001:2000 |
---|---|
Content And Storage | 2° to 8°C, away from light |
Form | Liquid |
Sterilization Method | 0.1 μm Sterile-filtered |
Product Code | Brand | Quantity | Shelf Life | With Additives | Without Additives | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Shelf Life | With Additives | Without Additives | Price | Quantity & Availability | ||
10673313
|
Cytiva
SH30284.01 |
500 mL | 12 Months | Glutamine, Ribonucleosides, Deoxyribonucleosides | No Phenol Red, No Sodium Pyruvate |
23.55 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10402952
|
Cytiva
SH30081.02 |
1000 mL | 18 Months | Glucose | No Glutamine, No Sodium Pyruvate |
23.20 €
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11988075
|
Cytiva
SH30081.FS |
6 x 500 mL | 18 Months | Glucose | No Glutamine, No Sodium Pyruvate |
143.00 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10544144
|
Cytiva
SH30081.LS |
6 x 1000 mL | 12 Months | Glucose | No Glutamine, No Sodium Pyruvate |
188.00 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11511871
|
Cytiva
SH30243.02 |
1000 mL | 12 Months | Glutamine, Sodium Pyruvate |
39.35 €
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10196223
|
Cytiva
SH30243.LS |
6 x 1000 mL | 12 Months | Glutamine, Sodium Pyruvate |
188.00 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10443142
|
Cytiva
SH30285.01 |
500 mL | 18 Months | Glucose, Sodium Pyruvate | No Glutamine |
23.70 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10524684
|
Cytiva
SH30243.FS |
6 x 500 mL | 12 Months | Glutamine, Sodium Pyruvate | No HEPES |
143.00 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10346712
|
Cytiva
SH30285.02 |
1000 mL | 18 Months | Glucose, Sodium Pyruvate | No Glutamine |
32.70 €
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10004873
|
Cytiva
SH30285.LS |
6 x 1000 mL | 18 Months | Glucose, Sodium Pyruvate | No Glutamine | N/A | |||
10359583
|
Cytiva
SH30285.FS |
6 x 500 mL | 18 Months | Glucose, Sodium Pyruvate | No Glutamine | N/A | |||
10523344
|
Cytiva
SH30604.01 |
500 mL | 18 Months | Glucose, Sodium Pyruvate | No Glutamine, No Phenol Red | N/A | |||
11551841
|
Cytiva
SH30081.01 |
500 mL | 18 Months | Glucose | No Glutamine, No Sodium Pyruvate |
24.70 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10658364
|
Cytiva
SH30585.02 |
1000 mL | 12 Months | Glucose | No Glutamine, No Phenol Red, No Sodium Pyruvate |
32.30 €
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10749864
|
Cytiva
SH30243.01 |
500 mL | 12 Months | Glutamine, Sodium Pyruvate | No HEPES |
24.70 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10294972
|
Cytiva
SH30284.02 |
1000 mL | 12 Months | Glutamine, Glucose | No Phenol Red, No Sodium Pyruvate |
32.40 €
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Format: Liquid
Specifications
ISO 9001:2000 | |
2° to 8°C, away from light | |
Liquid | |
0.1 μm Sterile-filtered |
Safety and Handling
missing translation for 'shelfLife' : 12 Months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title