Learn More
Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Powder
Brand:  Cytiva SH30013.02
22.04 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
 View more versions of this product
Product Code. 11511801
  • 23.25 € / Pack of 2
Explore more special offers
Description
Description
Buffer cell culture with HyClone™ Dulbecco's Phosphate Buffered Saline (PBS), Powder.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Powder
No Calcium, No Magnesium
2° to 8° C
Non-sterile
2 x 5 L