missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Solution
Buffer cell culture with PBS in solution.
Brand: Cytiva SH30264.FS
This item is not returnable.
View return policy
Description
- Format: Liquid
Specifications
Manufactured under cGMP and ISO 9001 certified facilities | |
HyClone Dulbecco's Phosphate Buffered Salines (DPBS) With Calcium, Magnesium, without Phenol Red | |
Liquid | |
Sterile | |
Calcium, Magnesium | |
≤0.025EU/mL | |
276±10mOsm/kg H2O | |
6 x 500 mL |
15° to 30°C | |
1 X | |
Dulbecco's Phosphate Buffered Saline | |
0.1 μm Sterile-filtered | |
No Phenol Red | |
Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time | |
7 to 7.6 | |
RUO |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction